Learn More
CPC Scientific H-Asp-Pro-Met-Ser-Ser-Thr-Tyr-Ile-Glu-Glu-Leu-Gly-Lys-Arg-Glu-Val-Thr-Ile-Pro-Pro-Lys-Tyr-Arg-Glu-Leu-Leu-Ala-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific CASP019A
Encompass
Selective inhibitor of Calpain I and II that exhibits no effect on papain or trypsin activity.ONE-LETTER SEQUENCE: DPMSSTYIEELGKREVTIPPKYRELLAMOLECULAR FORMULA: C140H227N35O44S1MOLECULAR WEIGHT:3136.57STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [128578-18-7], RESEARCH AREA: ApoptosisREFERENCES: M. Maki et al., JBC, 264, 18866 (1989)
Catalog No. 50-210-8955