Learn More
Medchemexpress LLC Calpain inhibitor VI (SJA6017) | 190274-53-4 | MFCD00954660 | 99.9% | 372.45 g·mol⁻¹ | C17H25FN2O4S | 1 MG
SDP
Catalog No. 5000307531
Encompass_Preferred
Calpain Inhibitor VI is a synthesized peptide aldehyde that reversibly inhibits calpain proteases and is provided for laboratory research use. It inhibits purified m-calpain with an IC50 of 80 nM, is supplied as a white to off-white solid of high purity, and is intended for biochemical and cellular studies of protease activity.
  • Peptide aldehyde calpain inhibitor.
  • Potent inhibition of m-calpain (IC50 80 nM).
  • Supplied as a white to off-white solid with high purity.
  • Suitable for biochemical assays and cellular research.
  • Recommended sealed storage away from moisture and light; in solvent store at -80°C up to 6 months.
Catalog No. 50-003-07531 Supplier Medchemexpress LLC Supplier No. HY1189331MG
Calpain Inhibitor VI is a synthesized peptide aldehyde that reversibly inhibits calpain proteases and is provided for laboratory research use. It inhibits purified m-calpain with an IC50 of 80 nM, is supplied as a white to off-white solid of high purity, and is intended for biochemical and cellular studies of protease activity.
  • Peptide aldehyde calpain inhibitor.
  • Potent inhibition of m-calpain (IC50 80 nM).
  • Supplied as a white to off-white solid with high purity.
  • Suitable for biochemical assays and cellular research.
  • Recommended sealed storage away from moisture and light; in solvent store at -80°C up to 6 months.