Learn More
Cayman Chemical Calpain Inhibitor I, Purity: ≥95%, CAS: 110044-82-1, 25 mg
SDP
Catalog No. 502969028
Encompass_Preferred
A synthetic tripeptide aldehyde that acts as a potent inhibitor of cysteine proteases including calpain I (Ki = 190 nM), calpain II (Ki = 220 nM) cathepsin B (Ki = 150 nM), and cathepsin L (Ki = 500 pM); prevents the degradation of IκBα and IκBβ by the ubiquitin-proteasome complex, which blocks activation of NFκB and the production of TNF and IL-1β
Catalog No. 50-296-9028 Supplier Cayman Chemical Supplier No. 1492125 MG
A synthetic tripeptide aldehyde that acts as a potent inhibitor of cysteine proteases including calpain I (Ki = 190 nM), calpain II (Ki = 220 nM) cathepsin B (Ki = 150 nM), and cathepsin L (Ki = 500 pM); prevents the degradation of IκBα and IκBβ by the ubiquitin-proteasome complex, which blocks activation of NFκB and the production of TNF and IL-1β