Learn More
Cayman Chemical Calpain Inhibitor II, Purity: ≥98%, CAS: 110115-07-6, 25 mg
SDP
Catalog No. 502968812
Encompass_Preferred
A cell permeable, peptide aldehyde inhibitor of calpain I (Ki = 120 nM), calpain II (Ki = 230 nM), cathepsin B (Ki = 100 nM), and cathepsin L (Ki = 0.6 nM)
Catalog No. 50-296-8812 Supplier Cayman Chemical Supplier No. 1599425 MG
A cell permeable, peptide aldehyde inhibitor of calpain I (Ki = 120 nM), calpain II (Ki = 230 nM), cathepsin B (Ki = 100 nM), and cathepsin L (Ki = 0.6 nM)