Learn More
Cayman Chemical CalpaIn InhIbItr II 5mg
SDP
Catalog No. 502973164
Encompass_Preferred
A cell permeable, peptide aldehyde inhibitor of calpain I (Ki = 120 nM), calpain II (Ki = 230 nM), cathepsin B (Ki = 100 nM), and cathepsin L (Ki = 0.6 nM)
Catalog No. 50-297-3164 Supplier Cayman Chemical Supplier No. 159945 MG
A cell permeable, peptide aldehyde inhibitor of calpain I (Ki = 120 nM), calpain II (Ki = 230 nM), cathepsin B (Ki = 100 nM), and cathepsin L (Ki = 0.6 nM)