Learn More
Cayman Chemical Calpain Inhibitor III, Purity: ≥95%, CAS: 88191-84-8, 10 mg
SDP
Catalog No. 502953961
Encompass_Preferred
A cell permeable, selective inhibitor of μ-calpain (calpain-1) and m-calpain (calpain-2); crosses the blood-brain barrier to inhibit brain cysteine protease activity; reported to have protective effects in numerous rodent models of neurotrauma and cardiac ischemia
Catalog No. 50-295-3961 Supplier Cayman Chemical Supplier No. 1428310 MG
A cell permeable, selective inhibitor of μ-calpain (calpain-1) and m-calpain (calpain-2); crosses the blood-brain barrier to inhibit brain cysteine protease activity; reported to have protective effects in numerous rodent models of neurotrauma and cardiac ischemia