Learn More
Cayman Chemical Calpain Inhibitor VI, Purity: ≥95%, CAS: 190274-53-4, 1 mg
SDP
Catalog No. 502995321
Encompass_Preferred
A calpain and cathepsin inhibitor; inhibits µ-calpain and m-calpain (IC50s = 7.5 and 78 nM, respectively); inhibits cathepsin B and cathepsin L (IC50s = 15 and 1.6 nM, respectively); selective over other cysteine and serine proteases and several enzymes; effective in in vitro (0.8 µM) and in vivo (100 mg/kg per day for 4 days in rats) models of cataract
Catalog No. 50-299-5321 Supplier Cayman Chemical Supplier No. 239421 MG
A calpain and cathepsin inhibitor; inhibits µ-calpain and m-calpain (IC50s = 7.5 and 78 nM, respectively); inhibits cathepsin B and cathepsin L (IC50s = 15 and 1.6 nM, respectively); selective over other cysteine and serine proteases and several enzymes; effective in in vitro (0.8 µM) and in vivo (100 mg/kg per day for 4 days in rats) models of cataract