Learn More
Cayman Chemical CalpaIn InhIbItr XII 10mg
SDP
Catalog No. 502970414
Encompass_Preferred
A reversible and selective inhibitor of calpain I (μ-calpain, Ki = 19 nM), with lower affinities for calpain II (m-calpain, Ki = 120 nM) and cathepsin B (Ki = 750 nM)
Catalog No. 50-297-0414 Supplier Cayman Chemical Supplier No. 1446610 MG
A reversible and selective inhibitor of calpain I (μ-calpain, Ki = 19 nM), with lower affinities for calpain II (m-calpain, Ki = 120 nM) and cathepsin B (Ki = 750 nM)