Learn More
Cayman Chemical Calpain Inhibitor XII, Purity: ≥90%, CAS: 181769-57-3, 1 mg
SDP
Catalog No. 502970120
Encompass_Preferred
A reversible and selective inhibitor of calpain I (μ-calpain, Ki = 19 nM), with lower affinities for calpain II (m-calpain, Ki = 120 nM) and cathepsin B (Ki = 750 nM)
Catalog No. 50-297-0120 Supplier Cayman Chemical Supplier No. 144661 MG
A reversible and selective inhibitor of calpain I (μ-calpain, Ki = 19 nM), with lower affinities for calpain II (m-calpain, Ki = 120 nM) and cathepsin B (Ki = 750 nM)