Learn More
CPC Scientific H-Glu(EDANS)-Pro-Leu-Phe-Ala-Glu-Arg-Lys(Dabcyl)-OH (trifluoroacetate salt) 5MG
SDP
Catalog No. 502108928
Encompass
An internally quenched peptide substrate for calpain-1 with optimal cleavage motifs flanking the scissile bondSEQUENCE: H-Glu(EDANS)-Pro-Leu-Phe-Ala-Glu-Arg-Lys(Dabcyl)-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: Glu(EDANS)-PLFAER-Lys(Dabcyl)MOLECULAR FORMULA: C72H97N17O16S1MOLECULAR WEIGHT: 1488.73STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [1914987-47-5]SYNONYMS: calpain-1 substrateRESEARCH AREA: CancerREFERENCES:D.Cuerrier et al., J. Biol. Chem., 280, 40632 (2005)SKU(s): CASB-001A, CASB-001B
Catalog No. 50-210-8928 Supplier CPC Scientific Supplier No. CASB001B
An internally quenched peptide substrate for calpain-1 with optimal cleavage motifs flanking the scissile bondSEQUENCE: H-Glu(EDANS)-Pro-Leu-Phe-Ala-Glu-Arg-Lys(Dabcyl)-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: Glu(EDANS)-PLFAER-Lys(Dabcyl)MOLECULAR FORMULA: C72H97N17O16S1MOLECULAR WEIGHT: 1488.73STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [1914987-47-5]SYNONYMS: calpain-1 substrateRESEARCH AREA: CancerREFERENCES:D.Cuerrier et al., J. Biol. Chem., 280, 40632 (2005)SKU(s): CASB-001A, CASB-001B