Description
- Calibration Weight Sets Adjusted to Meet or Exceed ASTM Class 1-4 Tolerances
- Troemner ASTM Class 1-4 offer high accuracy with tight tolerances and uncertainties of less than 0.25 of the tolerance
- Weight surfaces meet or exceed ASTM and have a highly polished finish
- Available in high grade stainless steels including Troemner Alloy 8 which possesses closely controlled density, extremely low magnetic properties, good stability, and resistance to corrosion
- Available with certificate options traceable to NIST, including ISO/IEC 17025 accredited certification or traceable certification to aid in meeting SOP and regulatory requirements
Specifications
Specifications
| Product Type | Calibration Weight |
| Material | Stainless Steel |
| Weight (Metric) | 100g-5g |
| Tolerance | Uncertainties of less than 1/4 of the tolerance. |