Learn More
Selleck Chemical LLC Calpeptin 10mg 117591-20-5
SDP
Catalog No. 501364734
Encompass_Preferred
Calpeptin is a potent, cell-permeable calpain inhibitor with ID50 of 52 nM, 34 nM, 138 nM, and 40 nM for Calpain I (porcine erythrocytes), Calpain II (porcine kidney), Papainb, and Calpain I (human platelets), respectively. Calpeptin attenuates apoptosis and intracellular inflammatory changes in muscle cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4734 Supplier Selleck Chemical LLC Supplier No. S739610MG
Calpeptin is a potent, cell-permeable calpain inhibitor with ID50 of 52 nM, 34 nM, 138 nM, and 40 nM for Calpain I (porcine erythrocytes), Calpain II (porcine kidney), Papainb, and Calpain I (human platelets), respectively. Calpeptin attenuates apoptosis and intracellular inflammatory changes in muscle cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.