Learn More
Medchemexpress LLC Calpeptin | 117591-20-5 | MFCD00155623 | 99.2% | 362.46 | C20H30N2O4 | 25 MG
SDP
Catalog No. 5000215509
Encompass_Preferred
Calpeptin is a cell-permeable peptide aldehyde used as a research reagent to inhibit calpain proteases in biochemical and cell-based studies. It shows potent activity against calpain I (ID50 ≈ 40 nM in human platelets) and also inhibits cathepsin K. The compound is supplied as a high-purity white to off-white solid with defined solubility and storage recommendations for laboratory use.
  • Potent calpain I inhibitor with ID50 around 40 nM.
  • Cell-permeable, suitable for cell-based assays.
  • High purity solid for analytical and biochemical work.
  • Soluble in DMSO at high concentrations for in vitro use.
  • Stable under recommended cold storage conditions as a powder.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-15509 Supplier Medchemexpress LLC Supplier No. HY10022325MG
Calpeptin is a cell-permeable peptide aldehyde used as a research reagent to inhibit calpain proteases in biochemical and cell-based studies. It shows potent activity against calpain I (ID50 ≈ 40 nM in human platelets) and also inhibits cathepsin K. The compound is supplied as a high-purity white to off-white solid with defined solubility and storage recommendations for laboratory use.
  • Potent calpain I inhibitor with ID50 around 40 nM.
  • Cell-permeable, suitable for cell-based assays.
  • High purity solid for analytical and biochemical work.
  • Soluble in DMSO at high concentrations for in vitro use.
  • Stable under recommended cold storage conditions as a powder.
  • For research use only; not for human or clinical use.