Learn More
Apexbio Technology LLC Calpeptin 117591-20-5 10mM (in 1mL Ethanol)
SDP
Catalog No. 5000061853
Encompass_Preferred
Calpeptin (CAS 117591-20-5) is a potent inhibitor of calpains calcium-dependent intracellular cysteine proteases involved in cellular differentiation proliferation and apoptosis It inhibits human calpain 1 with an IC50 of approximately 5 nM In vitro studies demonstrate that Calpeptin suppresses the production of TGF- 1 IL-6 and angiopoietin-1 as well as collagen synthesis in lung fibroblasts In mouse models of bleomycin-induced pulmonary fibrosis Calpeptin treatment reduces pulmonary tissue fibrosis accompanied by decreased expression of IL-6 TGF- 1 angiopoietin-1 and collagen type Ia1 mRNA Calpeptin serves as a valuable tool in fibrosis-related research
Catalog No. 50-000-61853 Supplier Apexbio Technology LLC Supplier No. A44115.1
Calpeptin (CAS 117591-20-5) is a potent inhibitor of calpains calcium-dependent intracellular cysteine proteases involved in cellular differentiation proliferation and apoptosis It inhibits human calpain 1 with an IC50 of approximately 5 nM In vitro studies demonstrate that Calpeptin suppresses the production of TGF- 1 IL-6 and angiopoietin-1 as well as collagen synthesis in lung fibroblasts In mouse models of bleomycin-induced pulmonary fibrosis Calpeptin treatment reduces pulmonary tissue fibrosis accompanied by decreased expression of IL-6 TGF- 1 angiopoietin-1 and collagen type Ia1 mRNA Calpeptin serves as a valuable tool in fibrosis-related research