Learn More
Takara Bio CalPhos™ Mammalian Transfection Kit, 700 Rxns
SDP
Supplier:  Takara Bio 631312
Encompass
The CalPhos Mammalian Transfection Kit is a complete calcium phosphate-based kit for the transfection of mammalian tissue culture cells. The kit contains reagents necessary to perform 100 transfections in 10 cm plates or 700 transfections (6-well plate format). 700 Rxns
Catalog No. NC9567834