Learn More
Medchemexpress LLC Calphostin C | 121263-19-2 | MFCD00133155 | 99.9% | 790.76 g·mol⁻1 | C44H38O14 | 100 UG
SDP
Catalog No. 5000244163
Encompass_Preferred
Calphostin C is a small-molecule, photo-dependent protein kinase C (PKC) inhibitor used in biochemical and cell biology research to probe PKC-mediated signaling and apoptosis. It has chemical formula C44H38O14 and a molecular weight of 790.76 g·mol⁻1. The compound is provided as a high-purity powder in 100 μg units; handle and store according to the supplier safety data sheet.
  • Potent, selective PKC inhibitor (IC50 = 0.05 μM).
  • Photo-dependent inhibition enabling light-controlled experiments.
  • High purity (>99.0%) powder for in vitro applications.
  • Chemical formula C44H38O14 and molecular weight 790.76 g·mol⁻1.
  • Supplied in 100 μg aliquots suitable for biochemical assays.
Catalog No. 50-002-44163 Supplier Medchemexpress LLC Supplier No. HY105416100UG
Calphostin C is a small-molecule, photo-dependent protein kinase C (PKC) inhibitor used in biochemical and cell biology research to probe PKC-mediated signaling and apoptosis. It has chemical formula C44H38O14 and a molecular weight of 790.76 g·mol⁻1. The compound is provided as a high-purity powder in 100 μg units; handle and store according to the supplier safety data sheet.
  • Potent, selective PKC inhibitor (IC50 = 0.05 μM).
  • Photo-dependent inhibition enabling light-controlled experiments.
  • High purity (>99.0%) powder for in vitro applications.
  • Chemical formula C44H38O14 and molecular weight 790.76 g·mol⁻1.
  • Supplied in 100 μg aliquots suitable for biochemical assays.