Learn More
Biocare Medical, LLC Calponin Antibody - Predilute, 6.0 ml
SDP
Supplier:  Biocare Medical, LLC PM172AA
Encompass_Preferred
Calponin a 34 kDa polypeptide, is a cytoskeleton-associated actin-binding protein that also interacts tropomyosin and calmodulin. Calponin antibody has been found to be useful as a marker for myoepithelial and basal lamina in differentiating microinvasive from in situ ductal carcinomas of the breast. Calponin antibody may also have applications in malignant myoepithelium and pleomorphic adenoma of salivary gland as well as a useful marker for fine needle aspirates of papillary breast lesions.
Catalog No. 50-830-90