Learn More
Aviva Systems Biology Corp CALR Antibody - middle region (ARP58880_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58880_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CALR. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human CALR. Alternate names: CRT, FLJ26680, RO, SSA, cC1qRApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-516