Learn More
Aviva Systems Biology Corp CALR Antibody - C-terminal region (ARP30114_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30114_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CALR. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human CALR. Alternate names: RO, CRT, SSA, cC1qRApplicationWB, IHC. Species reactivity: Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-54133