Learn More
Enzo Life Sciences Calreticulin monoclonal antibody (FMC 75) (50 µg)
SDP
Supplier:  Enzo Life Sciences ADISPA601D
Encompass_Preferred
Clone: FMC 75. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant calreticulin. UniProt: P27797 (human). Source: Purified from ascites. Species Reactivity: Human, Monkey. Applications: ICC, IHC (PS), IP, WB. Recommended dilutions: Immunoprecipitation (12.5µg/ml). Western Blot (1:1,000, ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-667-486