Learn More
Enzo Life Sciences Calreticulin polyclonal antibody (50 µg)
SDP
Supplier:  Enzo Life Sciences ADISPA600D
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the sequence near the C-terminus of human Calreticulin. UniProt: P27797. Source: Purified from rabbit serum. Species Reactivity: Human, Mouse, Rat, Dog, Bovine. Applications: ICC, IHC, IP, WB. Recommended dilutions: Western Blot (1:1,000, human; 1:500, mouse; ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-667-481