Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Calreticulin/CALR M 500ug
SDP

Catalog No. 5000195545
Encompass_Preferred

Recombinant GST-tagged mouse calreticulin (residues D18-L416) produced in E. coli and provided as purified protein for biochemical assays, antibody validation, and structural studies. The product is supplied at high purity and is intended for research use with recommended aliquoting for long-term storage.

  • Recombinant GST-tagged protein with N-terminal GST tag.
  • Expression system: E. coli for efficient production.
  • High purity greater than 95% as determined by reducing SDS-PAGE.
  • Sequence corresponds to accession P14211 (D18-L416).
  • Multiple pack sizes available, including a 500 μg option.
  • Store at -20°C; aliquot and store at -80°C for extended storage.

Catalog No. 50-001-95545 Supplier Medchemexpress LLC Supplier No. HYP72112500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant GST-tagged mouse calreticulin (residues D18-L416) produced in E. coli and provided as purified protein for biochemical assays, antibody validation, and structural studies. The product is supplied at high purity and is intended for research use with recommended aliquoting for long-term storage.

  • Recombinant GST-tagged protein with N-terminal GST tag.
  • Expression system: E. coli for efficient production.
  • High purity greater than 95% as determined by reducing SDS-PAGE.
  • Sequence corresponds to accession P14211 (D18-L416).
  • Multiple pack sizes available, including a 500 μg option.
  • Store at -20°C; aliquot and store at -80°C for extended storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.