Learn More
Medchemexpress LLC Calreticulin/CALR M 50ug
SDP
Catalog No. 5000195924
Encompass_Preferred
Recombinant mouse calreticulin (His-tagged), expressed in Pichia pastoris and supplied as a lyophilized powder formulated with trehalose for enhanced stability. The protein is suitable for biochemical and immunological assays where a high-quality antigen or binding partner is required; it features low endotoxin and high purity by reducing SDS-PAGE, and should be stored frozen for long-term stability.
  • Recombinant protein expressed in Pichia pastoris for eukaryotic folding and processing.
  • His-tag facilitates affinity purification and detection.
  • Lyophilized formulation with trehalose enhances long-term stability.
  • Low endotoxin level suitable for sensitive assays.
  • High purity by reducing SDS-PAGE for consistent experimental results.
  • Suitable for ELISA, western blot, antibody generation, and binding assays.
Catalog No. 50-001-95924 Supplier Medchemexpress LLC Supplier No. HYP7172150UG
Recombinant mouse calreticulin (His-tagged), expressed in Pichia pastoris and supplied as a lyophilized powder formulated with trehalose for enhanced stability. The protein is suitable for biochemical and immunological assays where a high-quality antigen or binding partner is required; it features low endotoxin and high purity by reducing SDS-PAGE, and should be stored frozen for long-term stability.
  • Recombinant protein expressed in Pichia pastoris for eukaryotic folding and processing.
  • His-tag facilitates affinity purification and detection.
  • Lyophilized formulation with trehalose enhances long-term stability.
  • Low endotoxin level suitable for sensitive assays.
  • High purity by reducing SDS-PAGE for consistent experimental results.
  • Suitable for ELISA, western blot, antibody generation, and binding assays.