Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Calreticulin/CALR M 50ug
SDP

Supplier:  Medchemexpress LLC HYP71721Y50UG

Encompass_Preferred

Recombinant N-terminal 6xHis-tagged mouse calreticulin expressed in Pichia pastoris and supplied as a 0.22 μm-filtered solution in PBS (pH 7.4) with 50% glycerol. Provided as research-grade purified protein with low endotoxin, it is suitable for biochemical assays, folding studies, and endoplasmic reticulum quality-control research.

  • N-terminal 6xHis-tagged recombinant mouse calreticulin.
  • Expressed in Pichia pastoris and supplied as a filtered solution.
  • Formulated in PBS pH 7.4 with 50% glycerol for stability.
  • Available in small research sizes including a 50 μg option.
  • Purity greater than 90% by reducing SDS-PAGE.
  • Molecular weight approximately 48.3 kDa; full-length mature protein.
  • Endotoxin <1 EU/μg (LAL), compatible with cell-based assays.
  • Store at -80°C; stable at -20°C for 3 months after opening; avoid repeated freeze-thaw.

Catalog No. 50-001-96078


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.