Learn More
Medchemexpress LLC Calreticulin/CALR M 5ug
SDP
Catalog No. 5000195926
Encompass_Preferred
Recombinant mouse calreticulin (CALR) produced in Pichia pastoris with an N-terminal 6xHis tag, supplied as a 5 μg solution. The protein is >90% pure by reducing SDS-PAGE and has an approximate molecular weight of 48.3 kDa. Supplied ready-to-use for biochemical and binding assays, structural studies, and as a control in cellular or immunological experiments; aliquot and store at -80°C and avoid repeated freeze-thaw cycles.
  • High purity suitable for biochemical and binding assays.
  • N-terminal 6xHis tag enables affinity purification and detection.
  • Produced in Pichia pastoris to support eukaryotic folding and post-translational modifications.
  • Supplied as a ready-to-use solution to simplify experiment setup.
  • Low-microgram amount appropriate for control experiments and pilot studies.
  • Stable when aliquoted and stored at -80°C to maintain activity.
Catalog No. 50-001-95926 Supplier Medchemexpress LLC Supplier No. HYP71721Y5UG
Recombinant mouse calreticulin (CALR) produced in Pichia pastoris with an N-terminal 6xHis tag, supplied as a 5 μg solution. The protein is >90% pure by reducing SDS-PAGE and has an approximate molecular weight of 48.3 kDa. Supplied ready-to-use for biochemical and binding assays, structural studies, and as a control in cellular or immunological experiments; aliquot and store at -80°C and avoid repeated freeze-thaw cycles.
  • High purity suitable for biochemical and binding assays.
  • N-terminal 6xHis tag enables affinity purification and detection.
  • Produced in Pichia pastoris to support eukaryotic folding and post-translational modifications.
  • Supplied as a ready-to-use solution to simplify experiment setup.
  • Low-microgram amount appropriate for control experiments and pilot studies.
  • Stable when aliquoted and stored at -80°C to maintain activity.