Learn More
AdipoGen anti-Calreticulin (human), mAb (CR213-2AG) (100 µg)
SDP
Supplier:  AdipoGen AG20A0079C100
Encompass_Preferred
Monoclonal Antibody. Recognizes human calreticulin. Isotype: Mouse IgG1kappa. Clone: CR213-2AG. Applications: ELISA, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. Calreticulin is involved in regulation of intracellular Ca2+ homoeostasis and ER Ca2+ capacity. It constitutes together with calnexin and ERp57 the ‘calreticulin/calnexin cycle’ that is responsible for folding and quality control of newly synthesized glycoproteins. Calreticulin has been implicated to play a role in many biological systems, including functions inside and outside the ER, indicating that the protein is a multi-process molecule.
Catalog No. 50-205-2881