Learn More
Enzo Life Sciences Calreticulin monoclonal antibody (FMC 75) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS6680200
Encompass_Preferred
Clone: FMC 75. Isotype: IgG1κ. Immunogen: Recombinant calreticulin. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human, Monkey. Applications: ICC, IHC (PS), IP, WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-201-3194