Learn More
Enzo Life Sciences Calreticulin monoclonal antibody (FMC 75) (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPA601F
Encompass_Preferred
Clone: FMC 75. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant calreticulin. Source: Purified from ascites. Species reactivity: Human, Monkey. Applications: ICC, IHC (PS), IP, WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7446