Learn More
Enzo Life Sciences Calreticulin monoclonal antibody (FMC 75) (DyLight 488 conjugate) (50 µg)
SDP
Supplier:  Enzo Life Sciences ADISPA601488D
Encompass_Preferred
Clone: FMC 75. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant calreticulin. UniProt: P27797 (human). Source: Purified from ascites. Species Reactivity: Human, Monkey. Applications: Flow Cytometry. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 0.09% sodium azide. Handling: Avoid freeze/thaw cycles. Protect from light.
Catalog No. 50-667-484