Learn More
Enzo Life Sciences Calreticulin monoclonal antibody (FMC 75) (PE conjugate) (50 µg)
SDP
Supplier:  Enzo Life Sciences ADISPA601PED
Encompass_Preferred
Clone: FMC 75. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant calreticulin. UniProt: P27797 (human). Source: Purified from ascites. Species Reactivity: Human, Monkey. Applications: Flow Cytometry. Recommended dilutions: Flow Cytometry (1:100). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 0.09% sodium azide. Handling: Avoid freeze/thaw cycles. Protect from light.
Catalog No. 50-667-488