Learn More
Enzo Life Sciences Calreticulin (NT) polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences ALX210170R100
Encompass_Preferred
Host: Chicken. Immunogen: Synthetic peptide corresponding to aa 24-43 (K24EQFLDGDAWTNRWVESKHK43) of mouse calreticulin. Species reactivity: Human, Mouse, Rat Hamster, Dog. Applications: ICC, WB. Recommended Dilutions: Western Blot (1:1,000) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Epitope-affinity purified IgY. Formulation: Liquid. Contains 1mg/ml BSA and 0.05% sodium azide.
Catalog No. 50-200-8084