Learn More
Enzo Life Sciences Calreticulin polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences ALX210126R100
Encompass_Preferred
Host: Rabbit. Immunogen: Recombinant human calreticulin. Species reactivity: Human, Mouse, Rat, Rabbit, Monkey, Dog. Applications: Flow Cytometry, ICC, IHC, IP, WB. Formulation: Liquid. Antiserum containing 0.05% sodium azide.
Catalog No. 50-200-8070