Learn More
Enzo Life Sciences Calreticulin (human), (recombinant) (His-tag) (50 µg)
SDP
Supplier:  Enzo Life Sciences ADISPP600D
Encompass_Preferred
Alternative name: CALR, CRT. MW: ~59kDa. Purity: ≥90% (SDS-PAGE; Western blot). Formulation: Liquid, In 50mM TRIS, pH 7.5, containing 150mM sodium chloride, 5mM calcium chloride and 5% glycerol. Source: Produced in E. coli. Human calreticulin is fused at the N-terminus to a His-tag. UniProt: P27797. Application: WB. Recommended dilutions: Western Blot (100 ng, control). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Long Term Storage: -80°C
Catalog No. 50-667-725