Learn More
Cayman Chemical CalretIculInhumnrecombIn 100ug
SDP
Catalog No. 502945270
Encompass_Preferred
Source: Recombinant human calreticulin expressed in HEK293 cells • Amino acids: 18-413 • MW: 45.99 kDa
Catalog No. 50-294-5270 Supplier Cayman Chemical Supplier No. 42565100 UG
Source: Recombinant human calreticulin expressed in HEK293 cells • Amino acids: 18-413 • MW: 45.99 kDa