Learn More
ABclonal Technology Calretinin Rabbit mAb
SDP
Catalog No. 5000007843
Encompass_Preferred
This gene encodes an intracellular calcium-binding protein belonging to the troponin C superfamily Members of this protein family have six EF-hand domains which bind calcium This protein plays a role in diverse cellular functions including message targeting and intracellular calcium buffering It also functions as a modulator of neuronal excitability and is a diagnostic marker for some human diseases including Hirschsprung disease and some cancers Alternative splicing results in multiple transcript variants
Catalog No. 50-000-07843 Supplier ABclonal Technology Supplier No. A2201020UL
This gene encodes an intracellular calcium-binding protein belonging to the troponin C superfamily Members of this protein family have six EF-hand domains which bind calcium This protein plays a role in diverse cellular functions including message targeting and intracellular calcium buffering It also functions as a modulator of neuronal excitability and is a diagnostic marker for some human diseases including Hirschsprung disease and some cancers Alternative splicing results in multiple transcript variants