Learn More
ATCC Calu-1; Lung Carcinoma; Human (Homo sapiens)
Catalog No. 502384870
Encompass
Calu-1 is a cell line exhibiting epithelial morphology that was isolated from the lungs of a 47-year-old, White male patient with epidermoid carcinoma. This product has applications in cancer research and toxicology.
Catalog No. 50-238-4870 Supplier ATCC Supplier No. HTB54
Calu-1 is a cell line exhibiting epithelial morphology that was isolated from the lungs of a 47-year-old, White male patient with epidermoid carcinoma. This product has applications in cancer research and toxicology.