Learn More
ATCC Calu-3; Lung Adenocarcinoma; Human (Homo sapiens)
Catalog No. 502384278
Encompass
Calu-3 epithelial cells are isolated from lung tissue derived from a 25-year-old, White, male patient with lung adenocarcinoma who received prior therapy with cytoxan, bleomycin, and adriamycin. This cell line is valuable for SARS-CoV-2 propagation in vitro, is a suitable transfection host, and has applications in cancer and toxicology research.
Catalog No. 50-238-4278 Supplier ATCC Supplier No. HTB55
Calu-3 epithelial cells are isolated from lung tissue derived from a 25-year-old, White, male patient with lung adenocarcinoma who received prior therapy with cytoxan, bleomycin, and adriamycin. This cell line is valuable for SARS-CoV-2 propagation in vitro, is a suitable transfection host, and has applications in cancer and toxicology research.