Learn More
Med Vet International Calvenza-03 EIV/Ehv Vaccine, Killed Virus, 20mL, 10 Dose
Catalog No. 502828720
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Equine Rhinopneumonitis-Influenza Vaccine, including Ohio/2003, Killed Virus, horses 6 months or older (including pregnant mares). Preservative: Gentamicin. Meat withdrawal: 21 days.
Catalog No. 50-282-8720 Supplier Med Vet International Supplier No. RXCALVENZAEIVEHV
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Equine Rhinopneumonitis-Influenza Vaccine, including Ohio/2003, Killed Virus, horses 6 months or older (including pregnant mares). Preservative: Gentamicin. Meat withdrawal: 21 days.