Learn More
Hach Company CalVer 2 Calcium Indicator, 454 g
Supplier:  Hach Company 28101H
Encompass
For determination of calcium hardness by EDTA titration methods. Active ingredient: Hydroxy Naphthol Blue. 454 g bottle
Catalog No. 50-199-7431