Learn More
Hach Company CalVer 2 Calcium Indicator Powder Pillows, 50 mL sample, pk/100
Catalog No. 501994457 Shop All Hach Company Products
Encompass
Indicator for determination of Calcium by EDTA titration. Powder pillows for 50-mL samples. Pack of 100 powder pillows.
Catalog No. 50-199-4457 Supplier Hach Company Supplier No. 85299
Indicator for determination of Calcium by EDTA titration. Powder pillows for 50-mL samples. Pack of 100 powder pillows.