Learn More
Aviva Systems Biology Corp Caly Antibody - middle region (ARP47188_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47188_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Caly. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Rat. Alternate names: Drd1ip, Calcyon, CalyApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-288