Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Calycanthine | 595-05-1 | 100.0% | 346.47 g/mol | C22H26N4 | 10 MG
SDP

Catalog No. 5000289840
Encompass_Preferred

Calycanthine is a naturally occurring indole alkaloid isolated from plants in the Calycanthaceae/related genera. It is supplied as a high-purity research chemical for in vitro studies and is reported to be a central nervous system toxin; handle per safety guidance and consult the SDS for hazard information.

  • High purity: 99.98%.
  • CAS number: 595-05-1.
  • Chemical formula: C22H26N4; molecular weight: 346.47 g/mol.
  • Appearance: light yellow to yellow solid.
  • Solubility: DMSO 100 mg/mL (288.63 mM); ultrasonic assistance recommended.
  • Storage: 4°C, sealed, away from moisture and light; in solution -80°C (6 months), -20°C (1 month).
  • Intended for research use only; not for human or veterinary use.

Catalog No. 50-002-89840 Supplier Medchemexpress LLC Supplier No. HYN512110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Calycanthine is a naturally occurring indole alkaloid isolated from plants in the Calycanthaceae/related genera. It is supplied as a high-purity research chemical for in vitro studies and is reported to be a central nervous system toxin; handle per safety guidance and consult the SDS for hazard information.

  • High purity: 99.98%.
  • CAS number: 595-05-1.
  • Chemical formula: C22H26N4; molecular weight: 346.47 g/mol.
  • Appearance: light yellow to yellow solid.
  • Solubility: DMSO 100 mg/mL (288.63 mM); ultrasonic assistance recommended.
  • Storage: 4°C, sealed, away from moisture and light; in solution -80°C (6 months), -20°C (1 month).
  • Intended for research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.