Learn More
TARGETMOL CHEMICALS INC CALYCOSIN 25MG
SDP
Catalog No. 502580064
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Calycosin (Cyclosin) acts as a selective estrogen receptor modulator. Calycosin induces apoptosis in human ovarian cancer SKOV3 cells by activating caspases and Bcl-2 family proteins. Calycosin suppresses breast cancer cell growth via ER(beta)-dependent regulation of IGF-1R p38 MAPK and PI3K/Akt pathways. Calycosin has antineoplastic effects against osteosarcoma through inducing apoptosis showing in vitro and in vivo investigations. Calycosin was shown to induce angiogenesis in human umbilical vein endothelial cell cultures (HUVEC) in vitro. Calycosin may be an effective skin-lightening agent that regulates the expression of melanogenic enzymes. Calycosin exhibited tyrosinase inhibitory activity with an IC(50) value of 38.4 microM. purity: 99%
Catalog No. 50-258-0064 Supplier TARGETMOL CHEMICALS INC Supplier No. T392325MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Calycosin (Cyclosin) acts as a selective estrogen receptor modulator. Calycosin induces apoptosis in human ovarian cancer SKOV3 cells by activating caspases and Bcl-2 family proteins. Calycosin suppresses breast cancer cell growth via ER(beta)-dependent regulation of IGF-1R p38 MAPK and PI3K/Akt pathways. Calycosin has antineoplastic effects against osteosarcoma through inducing apoptosis showing in vitro and in vivo investigations. Calycosin was shown to induce angiogenesis in human umbilical vein endothelial cell cultures (HUVEC) in vitro. Calycosin may be an effective skin-lightening agent that regulates the expression of melanogenic enzymes. Calycosin exhibited tyrosinase inhibitory activity with an IC(50) value of 38.4 microM. purity: 99%