Learn More
Apexbio Technology LLC Calycosin-7-glucoside 20633-67-4 20mg
SDP
Catalog No. 5000087746
Encompass_Preferred
Calycosin-7-glucoside (CAS 20633-67-4) is a major isoflavonoid glycoside isolated from Astragalus membranaceus var mongholicus It is known to modulate multiple cellular signaling pathways including antioxidant and anti-inflammatory mechanisms by regulating reactive oxygen species and inhibiting pro-inflammatory mediators Research indicates that Calycosin-7-glucoside exhibits cytoprotective and immunomodulatory effects making it relevant for studies on oxidative stress inflammation and immune regulation The compound is widely used in biomedical investigation of botanical therapeutics and cellular defense processes
Catalog No. 50-000-87746 Supplier Apexbio Technology LLC Supplier No. N141320
Calycosin-7-glucoside (CAS 20633-67-4) is a major isoflavonoid glycoside isolated from Astragalus membranaceus var mongholicus It is known to modulate multiple cellular signaling pathways including antioxidant and anti-inflammatory mechanisms by regulating reactive oxygen species and inhibiting pro-inflammatory mediators Research indicates that Calycosin-7-glucoside exhibits cytoprotective and immunomodulatory effects making it relevant for studies on oxidative stress inflammation and immune regulation The compound is widely used in biomedical investigation of botanical therapeutics and cellular defense processes