Learn More
TARGETMOL CHEMICALS INC CALYCOSIN-7-O-BETA-D-GLU 10MG
SDP
Catalog No. 502580287
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Calycosin-7-O-(beta)-D-glucoside (calycosin-7-O-beta-D-glucopyranoside) possesses a protective effect on rat hepatocytes an inhibitory effect on COX-2 activity. Calycosin 7-O-(beta)-D-glucoside has antimicrobial activity. Calycosin 7-O-(beta)-D-glucoside is a promising anti-HIV agent. It shows scavenging activity to DPPH radicals and superoxide anion. Calycosin 7-O-(beta)-D-glucopyranoside possesses an alleviating effect on osteoarthritis and can increase the fluidity of brain cell membrane in ischemia-reperfusion rats. purity: 99%
Catalog No. 50-258-0287 Supplier TARGETMOL CHEMICALS INC Supplier No. T338810MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Calycosin-7-O-(beta)-D-glucoside (calycosin-7-O-beta-D-glucopyranoside) possesses a protective effect on rat hepatocytes an inhibitory effect on COX-2 activity. Calycosin 7-O-(beta)-D-glucoside has antimicrobial activity. Calycosin 7-O-(beta)-D-glucoside is a promising anti-HIV agent. It shows scavenging activity to DPPH radicals and superoxide anion. Calycosin 7-O-(beta)-D-glucopyranoside possesses an alleviating effect on osteoarthritis and can increase the fluidity of brain cell membrane in ischemia-reperfusion rats. purity: 99%