Learn More
Cayman Chemical CalycosIn 7-OD-GlucosIde 10mg
SDP
Catalog No. 502935306
Encompass_Preferred
An isoflavone glycoside; scavenges 34.5% of generated DPPH radicals compared to a vehicle control in a cell-free assay at 100 μg/ml; inhibits LPS-induced cytotoxicity, ROS production, and cytoskeletal defects in HUVECs at 10 μg/ml; reduces brain infarct volume in a rat model of ischemia-reperfusion injury induced by MCAO at 26.8 mg/kg
Catalog No. 50-293-5306 Supplier Cayman Chemical Supplier No. 2509510 MG
An isoflavone glycoside; scavenges 34.5% of generated DPPH radicals compared to a vehicle control in a cell-free assay at 100 μg/ml; inhibits LPS-induced cytotoxicity, ROS production, and cytoskeletal defects in HUVECs at 10 μg/ml; reduces brain infarct volume in a rat model of ischemia-reperfusion injury induced by MCAO at 26.8 mg/kg