Learn More
Enzo Life Sciences Calmodulin (bovine brain), (agarose immobilized) (1 ml)
SDP
Supplier:  Enzo Life Sciences BMLSE1020001
Encompass_Preferred
Alternative name: CaM, Phosphodiesterase 3':5'-cyclic nucleotide activator. Concentration: Calmodulin concentration is 1 mg/ml beads. Formulation: Liquid. Suspension. Source: Isolated from bovine brain. Calmodulin conjugated to 4% cross-linked agarose beads via a 1 atom hydrophilic spacer arm. Long Term Storage: +4°C.
Catalog No. 50-201-1925