Learn More
MSC Albion Caster Cam Brake Kit, 1/2 in. ID, 6 in. to 8 in. Wheel, 1 ea.
Catalog No. 5000516724
Encompass
This caster cam brake kit provides reliable locking functionality for compatible heavy-duty casters. Designed for side-pedal operation, the cam action mechanism securely engages the wheel to prevent unwanted movement, enhancing safety and stability in various industrial applications.
  • Features side-pedal cam action brake mechanism
  • Compatible with 6 in. and 8 in. casters with 2 in. wheel width
  • Suitable for use with series 12, 16, 61, and 62 casters
  • Designed for use with roller, oil, and oilex bearings
  • Compatible with both swivel and rigid caster configurations
  • Not compatible with precision or Delrin bearings
Catalog No. 50-005-16724 Supplier MSC Supplier No. 01859040
This caster cam brake kit provides reliable locking functionality for compatible heavy-duty casters. Designed for side-pedal operation, the cam action mechanism securely engages the wheel to prevent unwanted movement, enhancing safety and stability in various industrial applications.
  • Features side-pedal cam action brake mechanism
  • Compatible with 6 in. and 8 in. casters with 2 in. wheel width
  • Suitable for use with series 12, 16, 61, and 62 casters
  • Designed for use with roller, oil, and oilex bearings
  • Compatible with both swivel and rigid caster configurations
  • Not compatible with precision or Delrin bearings