Learn More
Grainger Smith Bearing Needle Roller Cam Follower, 1-1/2 in. Dia., Standard Stud, 1 ea.
Catalog No. 502770431
Encompass_Preferred
These standard duty, needle roller cam followers are engineered for general purpose applications where running and shock loads are moderate. Constructed from high-alloy bearing quality steel, these components feature a screwdriver slot stud design and are available in sealed versions.
  • Recommended for moderate running and shock loads
  • Manufactured from high-alloy bearing quality steel
  • Static radial load capacity of 5,730 lbf
  • Dynamic radial load capacity of 5,560 lbf
  • Operating temperature range of -30°F to 250°F
  • Standard stud type with slotted drive for installation
Catalog No. 50-277-0431 Supplier Grainger Supplier No. 13W670
These standard duty, needle roller cam followers are engineered for general purpose applications where running and shock loads are moderate. Constructed from high-alloy bearing quality steel, these components feature a screwdriver slot stud design and are available in sealed versions.
  • Recommended for moderate running and shock loads
  • Manufactured from high-alloy bearing quality steel
  • Static radial load capacity of 5,730 lbf
  • Dynamic radial load capacity of 5,560 lbf
  • Operating temperature range of -30°F to 250°F
  • Standard stud type with slotted drive for installation