Learn More
Grainger Smith Bearing Cam Follower, Needle Bearing, 1-1/8 in. Roller Diameter, 7/16-20 Thread, 1 ea.
Catalog No. 502769959
Encompass_Preferred
This standard cam follower is a needle roller bearing designed for applications where moderate running and shock loads are present. Constructed from standard bearing steels with a flat roller surface profile, this stud-type bearing offers reliable performance in demanding industrial environments. It features a relubricatable design and is packed with general-purpose EP grease for consistent operation.
  • Roller diameter of 1 1/8 in. and roller width of 5/8 in.
  • Dynamic load capacity of 3,030 lbf and static load capacity of 3,100 lbf.
  • Features a 7/16-20 thread size and a 1/4 in. hex socket drive.
  • Suitable for operating temperatures ranging from -30 °F to 250 °F.
  • Designed with a full complement, single-row needle bearing configuration.
  • Includes provisions for relubrication via 3/16 in. drive type fittings.
Catalog No. 50-276-9959 Supplier Grainger Supplier No. 13W710
This standard cam follower is a needle roller bearing designed for applications where moderate running and shock loads are present. Constructed from standard bearing steels with a flat roller surface profile, this stud-type bearing offers reliable performance in demanding industrial environments. It features a relubricatable design and is packed with general-purpose EP grease for consistent operation.
  • Roller diameter of 1 1/8 in. and roller width of 5/8 in.
  • Dynamic load capacity of 3,030 lbf and static load capacity of 3,100 lbf.
  • Features a 7/16-20 thread size and a 1/4 in. hex socket drive.
  • Suitable for operating temperatures ranging from -30 °F to 250 °F.
  • Designed with a full complement, single-row needle bearing configuration.
  • Includes provisions for relubrication via 3/16 in. drive type fittings.